0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
TISSUE MARKING DYE ORANGE,59ML 1 * 1 items
SHAN5000O
:
Epredia
New Product
TISSUE MARKING DYE ORANGE,59ML 1 * 1 items
SHAN5000O
:
Epredia
New Product