Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
TISSUE MARKING DYE BLUE ,59ML 1 * 1 items
TISSUE MARKING DYE BLUE ,59ML 1 * 1 items
Print
TISSUE MARKING DYE BLUE ,59ML 1 * 1 items
Catalog # SHAN5000BL
Supplier:
Epredia
TISSUE MARKING DYE BLUE ,59ML 1 * 1 items
Catalog # SHAN5000BL
Supplier:
Epredia
Expand image
Return to Top
TISSUE MARKING DYE BLUE ,59ML 1 * 1 items
Add to cart
Price
48,90 €
Log in
for availability
Unit of Measure
1 * 1 items
-
+
Add to cart
Return Policy
Product Details & Documents