Aspartame
Poly bottle, 2,5 kg
Catalog # SPCMA1378-2.5KG
Supplier: Spectrum Chemical
Jump to:
Product Details & Documents
Aspartame, Powder, NF. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).
- Transmittance at 430 nm: minimum 0,95
- Specific rotation (A)D: +14,5 to +16,5
- Loss On drying: Max. 4,5%
- Residue on ignition: Max. 0,2%
Specifications
Specifications
- Pack type:Poly bottle
- Pk:2,5 kg
- Cat. No.:SPCMA1378-2.5KG
- Molecular formula:C₁₄H₁₈N₂O₅
- Form:Powder
- Protein/peptide name:Aspartame
- Storage conditions:Ambient
- Protein synonyms:Methyl aspartylphenylalanate|stereoisomer|L-Phenylalanine, N-L-alpha-aspartyl, 1-methyl ester|N-L-alpha Aspartyl-L-phenlalanine 1-methyl ester|Succinamic acid|Nutrasweet|N-methyl ester|3-amino-N-(alpha-carboxyphenethyl)-|1-Methyl N-L-alpha-aspartyl-L-phenylalanine|Aspartylphenylalanine methyl ester|3-Amino-N-(alpha-carboxyphenethyl) succinamic acid N-methyl ester
- Purity:98,0% (Dried basis)
- MDL No.:MFCD00002724
- Grade:NF
- Molecular weight:294,30 g/mol
- CAS No.:22839-47-0



