Orders LinkContactUsLinkComponent
Alpha Sterile Kits
White, M, 40/pack, +
Catalog # ISOF80551M
Supplier:   ISOFIELD IRELAND
Alpha Sterile Kits
Catalog # ISOF80551M
Supplier:  ISOFIELD IRELAND
Alpha Sterile Kits
Alpha Sterile Kits
White, M, 40/pack, +
Price
784,00 €
Log in for availability
1 * 40 items
Product Details & Documents
Sterile coverall with mandarin collar, hood, and overboots.
  • Manufactured in a facility operating under ISO 9001:2015 quality management system
  • Processed in a NEBB certified ISO Class 5 cleanroom
Mandarin collar coverall:
GMP Grade A and B, ISO Class 4 and 5 cleanroom compatible
Two-layer mandarin collar provides secure fit at neck area
Zip flap with double-sided tape prevents ingress and egress of particles
Low-linting, anti-static fabric suitable for chemo handling
Elasticated thumb loops
Back elastic to prevent bellows effect
Hood:
Latex-free enclosed elastic face opening for comfortable fit
Extra-long yoke for added protection
Three panel hood design to minimise skin exposure
Overboots:
Extra-long knee height
Two-ties for added security
Anti-slip PU sole
 : 1 set (hood/coverall/boots) per sealed inner bag. 1 inner bag per sealed outer bag. 40 outer bags per carton liner with twist tie (S to XXL). 35 sets per carton (3XL).
Specifications
  • Clothing Size:
    M
  • Packed:
    40/pack
  • Sterile:
    +
  • Colour:
    White
  • Seq:
    02
  • Cat. No.:
    ISOF80551M
  • Cleanroom class:
    10/ISO 4
  • Anti-static:
    +
  • Closure type:
    Zip closure