Disposable cannulas
Disposable cannulas, G14, Green, 100 mm, +
Catalog # EHRS15886510C
Supplier: EHRHARDT SOEHNE
Jump to:
Product Details & Documents
Disposable cannulas.
- Pyrogen-free, not toxic
- With lancet cut
- EO-sterilisation
Documents
Certificates
Search by Lot NumberSpecifications
- Gauge:G14
- Sterile:+
- Colour:Green
- Description:Disposable cannulas
- Length:100 mm
- Seq:24