Mixing Systems, SpinBag
SpinBag mixing systems, 10 L
Catalog # PARKPHSBU010LGI
Supplier: PARKER HANNIFIN
Jump to:
Product Details & Documents
Parker's compact SpinBag Mixing System offers the advantages of single-use in a bench top mixing solution.
- Constructed of fully validated DuraPure C93 film offering added seal strength, low gas permeability and optical clarity which can be heated to 60 °C
- Stores flat and ideal for preparation of cell culture media, buffers, bioreactor feeds, and suspension/re-suspension applications
- Easily integrated with filters, sensors and pumping systems
- Available with standard configurations, or a variety of ports and tubing options in non-irradiated or gamma irradiated
- Platinum-cured silicone coated magnetic stir bar offers optimal mixing performance and eliminates the need to validate another material
- Manufactured in an ISO Class 7 cleanroom
Parker's 3D SpinBag bioprocess containers are constructed of durable, particle-free, DuraPure C93 film, each containing a platinum-cured silicone coated magnetic spin bar. When placed on a mixing plate, the spin bar rotates to create a vortex of motion for thorough mixing/stirring of fluid contents. Parker SpinBags are available in a variety of sizes and port configurations to easily integrate within your application.
Specifications
- Description:SpinBag mixing systems, 10 L
- Seq:01
Accessories
Product Information
- Catalog #AvailabilityPrice
- SpinBag housing only, 50 L, polycarbonateSpinBag housing only, 50 L, polycarbonatePARKPHSBH050LLog in for availability1 * 1 SET - 822,00 €
Product Information
- Catalog #AvailabilityPrice
- SpinBag housing only, 15 L, polycarbonateSpinBag housing only, 15 L, polycarbonatePARKPHSBH015LLog in for availability1 * 1 SET - 686,00 €



