0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
Standards
Colour Reference and Spectroscopy Standards
Colour Standards ASTM
Colour Standards ASTM
ASTM 7 colour standard, single solution, 500 mL
PROOALK-ASTM70
:
LGC Standards PROMOCHEM
Colour Standards ASTM
PROOALK-ASTM70
:
LGC Standards PROMOCHEM
:
Colour Standards ASTM
ASTM 7 colour standard, single solution, 500 mL
:
288,00 €
1 * 500 mL
-
+
Pk:
500 mL
Description:
ASTM 7 colour standard, single solution
Cat. No.:
PROOALK-ASTM70
Storage temperature:
RT
Method:
Colour analysis