Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Recombinant rabbit monoclonal [RM274] antibody to mTOR. 1 * 50 µl
Catalog # ANTIA121387-50
Supplier:
Antibodies.com
New Product
Expand image
Recombinant rabbit monoclonal [RM274] antibody to mTOR. 1 * 50 µl
Catalog # ANTIA121387-50
Supplier:
Antibodies.com
New Product