0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
Recombinant goat monoclonal [RMG03] antibody to Rabbit IgG (Biotin). 1 * 25 µG
ANTIA121343-25
:
Antibodies.com
New Product
Recombinant goat monoclonal [RMG03] antibody to Rabbit IgG (Biotin). 1 * 25 µG
ANTIA121343-25
:
Antibodies.com
New Product