Orders LinkContactUsLinkComponent
Cellulose Extraction Thimbles, C300
25×70 mm, 1,3 mm
  SARTFT-1201-025070
   Low Manufacturing Impact
 :   Sartorius
Cellulose Extraction Thimbles, C300
  SARTFT-1201-025070
 :  Sartorius
   Low Manufacturing Impact
Cellulose Extraction Thimbles, C300
Cellulose Extraction Thimbles, C300
25×70 mm, 1,3 mm
117,00 €
1 * 25 items
These cellulose thimbles are primarily designed to fit the Soxhlet apparatus and potential applications include environmental monitoring (e.g. separation of dust, aerosols, gas or air streams) and food control (e.g. extraction of fats, emulsifiers or additives).
  • Composed of high-purity cellulose for uncontaminated results
    Engineered for superior mechanical stability to withstand rigorous use
    Designed for a precise fit across a diverse range of extractors
    Exhibits outstanding chemical resistance, ensuring reliability in various extraction processes
They are free of binders and offer a high purity which ensures consistent and high overall flow rates.
Typical wall thickness are 1,3 mm for inner diameters lower than 35 mm and 1,7 mm for inner diameters higher than 35 mm.
  • Int.Ø×H:
    25×70 mm
  • Seq:
    5
  • Thickness:
    1,3 mm
  • Cat. No.:
    SARTFT-1201-025070
  • Environmental certification:
    Renewable Energy Certificate
  • Material:
    Cellulose
  • Grade:
    C300
  • Environmentally Preferable:
    Low Manufacturing Impact