Order Entry
Export
Orders LinkContactUsLinkComponent
XFD488 NHS Ester fluorescent probe

XFD488 NHS Ester fluorescent probe

Plastic vial, 5 mg
Catalog # AATB71902
Irritant
Supplier:   AAT Bioquest
XFD488 NHS Ester fluorescent probe
Catalog # AATB71902
Supplier:  AAT Bioquest
CAS Number:  
XFD488 NHS Ester fluorescent probe
XFD488 NHS Ester fluorescent probe

Plastic vial, 5 mg

CAS Number:  
Price
1 100,00 €
Log in for availability
1 * 5 mg
Product Details & Documents

XFD488 NHS Ester (Succinimidyl Ester) is the same molecule to the Alexa Fluor® 488 NHS Ester (Alexa Fluor® is the trademark of ThermoFisher).

  • pH-insensitive
  • High photostability
  • Brighter signal

Caution: For research use only.

  • Molecular Weight:643.4 g/mol
  • ADR:5,II
Specifications

Specifications

  • Pack type:
    Plastic vial
  • Pk:
    5 mg

Specifications