Orders LinkContactUsLinkComponent
ReadiUse™ 1 Kb Plus DNA Ladder
ReadiUse™ 1 Kb Plus DNA ladder, 100 µl
Catalog # AATB60050
Supplier:   AAT Bioquest
ReadiUse™ 1 Kb Plus DNA Ladder
Catalog # AATB60050
Supplier:  AAT Bioquest
CAS Number:  
ReadiUse™ 1 Kb Plus DNA Ladder
ReadiUse™ 1 Kb Plus DNA Ladder
ReadiUse™ 1 Kb Plus DNA ladder, 100 µl
CAS Number:  
Price
142,00 €
Log in for availability
1 * 100 µl
Product Details & Documents
ReadiUse™ 1 kb Plus DNA ladder is a ready-to-use DNA ladder, which is pre-mixed with loading dye for direct gel loading.
  • Stable for 12 months
Quality tested in-house.
 : For research use only.
Specifications
Specifications
  • Pk:
    100 µl
  • Description:
    ReadiUse™ 1 Kb Plus DNA ladder
  • Cat. No.:
    AATB60050
Specifications