CytoWatch™ Multiplexing Buffer for Flow Cytometry, 20X
CytoWatch™ Multiplexing buffer for flow cytometry, 20X, 1.000 Tests
Catalog # AATB37011
Supplier: AAT Bioquest
CytoWatch™ Multiplexing Buffer for Flow Cytometry, 20X
Catalog # AATB37011
Supplier: AAT Bioquest
CAS Number:
Jump to:
Product Details & Documents
CytoWatch™ Multiplexing Buffer is a buffer optimised for the multicolor flow cytometric analysis of cells.
- Designed for ease of use
- Enhances multicolor flow experiments with two or more staining reagents
CytoWatch™ Multiplexing Buffer is a concentrated solution that is directly added to the staining solutions of certain fluorescent antibody conjugates or other fluorescent reagents before staining cells.
Caution: For research use only.
Specifications
Specifications
- Pk:1.000 Tests
- Description:CytoWatch™ Multiplexing buffer for flow cytometry, 20X