Orders LinkContactUsLinkComponent
CytoWatch™ Multiplexing Buffer for Flow Cytometry, 20X
CytoWatch™ Multiplexing Buffer for Flow Cytometry, 20X
CytoWatch™ Multiplexing buffer for flow cytometry, 20X, 100 Tests
Catalog # AATB37010
Supplier:   AAT Bioquest
CytoWatch™ Multiplexing Buffer for Flow Cytometry, 20X
Catalog # AATB37010
Supplier:  AAT Bioquest
CAS Number:  
CytoWatch™ Multiplexing Buffer for Flow Cytometry, 20X
CytoWatch™ Multiplexing Buffer for Flow Cytometry, 20X
CytoWatch™ Multiplexing buffer for flow cytometry, 20X, 100 Tests
CAS Number:  
Price
113,00 €
Log in for availability
1 * 100 Tests
Product Details & Documents
CytoWatch™ Multiplexing Buffer is a buffer optimised for the multicolor flow cytometric analysis of cells.
  • Designed for ease of use
  • Enhances multicolor flow experiments with two or more staining reagents
CytoWatch™ Multiplexing Buffer is a concentrated solution that is directly added to the staining solutions of certain fluorescent antibody conjugates or other fluorescent reagents before staining cells.
Caution: For research use only.
Specifications
Specifications
  • Pk:
    100 Tests
  • Description:
    CytoWatch™ Multiplexing buffer for flow cytometry, 20X
Specifications