Order Entry
Export
Orders LinkContactUsLinkComponent
Mesitylene ≥98.0%, GPR RECTAPUR®

Mesitylene ≥98.0%, GPR RECTAPUR®

Metal drum, 25 L
Catalog # 92512.460
Flammable
Health
Irritant
Environment
Supplier:   VWR Chemicals
Mesitylene ≥98.0%, GPR RECTAPUR®
Catalog # 92512.460
Supplier:  VWR Chemicals
CAS Number:  108-67-8
Mesitylene ≥98.0%, GPR RECTAPUR®

Metal drum, 25 L

CAS Number:  108-67-8
Price
Inquire For Price
Log in for availability
1 * 25 L
Product Details & Documents
Synonyms:  1,3,5-Trimethylbenzene
  • CAS Number:108-67-8
  • Formula:C₉H₁₂
  • MDL Number:MFCD00008538
  • Molecular Weight:120.19 g/mol
  • Merck Index:13,05934
  • Boiling Pt:165 °C (1013 hPa)
  • Melting Pt:–45 °C
  • Density:0.8637 g/cm³ (20 °C)
  • Flash Pt:48 °C
  • Storage Temperature:Ambient
  • EINECS:203-604-4
  • UN:2325
  • ADR:3,III
Specifications

Specification Test Results

  • Assay:
    ≥ 98,0 %
  • Identification (IR):
    Passes test
  • Density (20/4):
    0,864 - 0,866

Specifications

  • Pack type:
    Metal drum
  • Pk:
    25 L
  • Cat. No.:
    92512.460

Specifications