Mesitylene ≥98.0%, GPR RECTAPUR®
Glass bottle, 500 mL
Supplier: VWR Chemicals
Jump to:
Product Details & Documents
Synonyms: 1,3,5-Trimethylbenzene
- CAS Number:108-67-8
- Formula:C₉H₁₂
- MDL Number:MFCD00008538
- Molecular Weight:120.19 g/mol
- Merck Index:13,05934
- Boiling Pt:165 °C (1013 hPa)
- Melting Pt:–45 °C
- Density:0.8637 g/cm³ (20 °C)
- Flash Pt:48 °C
- Storage Temperature:Ambient
- EINECS:203-604-4
- UN:2325
- ADR:3,III
Documents
Specifications
Specification Test Results
- Assay:≥ 98,0 %
- Identification (IR):Passes test
- Density (20/4):0,864 - 0,866
Specifications
- Pack type:Glass bottle
- Pk:500 mL



