Order Entry
Export
Orders LinkContactUsLinkComponent
Human Recombinant IL-31

Human Recombinant IL-31

1.000 µG
Catalog # STMC78216.3
Human Recombinant IL-31
Catalog # STMC78216.3
CAS Number:  
Human Recombinant IL-31
Human Recombinant IL-31

1.000 µG

CAS Number:  
Price
7 220,00 €
Log in for availability
1 * 1.000 µG
Product Details & Documents

Interleukin 31 (IL-31), a four-helix bundle inflammatory cytokine, belongs to the IL-6 cytokine family which includes IL-6, oncostatin M, leukemia inhibitory factor (LIF), and cardiotrophin-1 ( Dillon et al.). IL-31 signals through a heterodimer composed of IL-31RA (also known as gp130-like receptor or GPL) and the oncostatin-M receptor (OSMR), both of which are expressed on monocytes (Diveu et al.; Ghilardi et al.), epithelial cells (Ip et al.), and keratinocytes (Kato et al.). Signaling through the GPL/OSMR complex activates the JAK/STAT, RAS/ERK, and PI3K/AKT signaling pathways, resulting in the downstream activation of STAT1, STAT3, and STAT5 transcription factors (Cornelissen et al.; Dambacher et al.; Dillon et al.; Ip et al.). IL-31 responses have been associated with allergic responses and inflammatory skin diseases including atopic dermatitis (Cornelissen et al.; Gangemi et al.).

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifications

Specifications

  • Pk:
    1.000 µG
  • Cat. No.:
    STMC78216.3
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    IL-31
  • Purity:
    ≥95%
  • Species:
    Human

Specifications