Order Entry
Export
Orders LinkContactUsLinkComponent
Human Recombinant IL-4, ACF

Human Recombinant IL-4, ACF

1.000 µG
Catalog # STMC78147.2
Human Recombinant IL-4, ACF
Catalog # STMC78147.2
CAS Number:  
Human Recombinant IL-4, ACF
Human Recombinant IL-4, ACF

1.000 µG

CAS Number:  
Price
4 840,00 €
Log in for availability
1 * 1.000 µG
Product Details & Documents

Human Interleukin 4 (IL-4) is important for immune responses to helminth infection as well as in allergic responses (Olpihant et al.). The IL-4 receptor consists of a heterodimer of IL-4Ra and common gamma chain. IL-4 receptor engagement leads to the activation of JAK1/3 and the recruitment of STAT6 and IRS1/2 (Nelms et al.). IL-4 drives immunoglobulin class switching in B cells (to IgE, IgG4), mast cell hyperplasia, mucus production, and the differentiation of naïve T cells into T helper type 2 (Th2) cells, which produce IL-4, IL-5, IL-10, and IL-13 (Bao et al.; Olpihant et al.; Nelms et al.). In addition to Th2 T cells, IL-4 is produced by CD4+ NK T cells, γ/δ T cells, activated basophils, eosinophils, and mast cells. IL-4 consists of 130 amino acids with a predicted molecular weight of 15.1 kDa. Human IL-4 does not cross-react with mouse cells (Park et al.). This product is animal component-free.

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifications

Specifications

  • Pk:
    1.000 µG
  • Cat. No.:
    STMC78147.2
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    IL-4, ACF
  • Purity:
    ≥95%
  • Species:
    Human

Specifications