Order Entry
Export
Orders LinkContactUsLinkComponent
Human Recombinant TNF-beta

Human Recombinant TNF-beta

50 µG
Catalog # STMC78125.1
Human Recombinant TNF-beta
Catalog # STMC78125.1
CAS Number:  
Human Recombinant TNF-beta
Human Recombinant TNF-beta

50 µG

CAS Number:  
Price
462,00 €
Log in for availability
1 * 50 µG
Product Details & Documents

Tumor necrosis factor-beta (TNF-β), also known as lymphotoxin-alpha, is a member of the TNF family. It is a pro-inflammatory cytokine that activates NF-kB, MAPK, and PI3K/AKT pathways upon binding to TNF receptors 1 and 2. It is produced mainly by T cells, though other cells can express TNF-β at lower levels (Chu). TNF-β is involved in autoimmune disorders, lymph node development, and mediating the inflammatory demyelination process (Seleznik et al.; McCarthy et al.).

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifications

Specifications

  • Pk:
    50 µG
  • Cat. No.:
    STMC78125.1
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    TNF-beta
  • Purity:
    ≥95%
  • Species:
    Human

Specifications