Bacillol® 30 Sensitive Green Tissues
Bacillol 30 sensitive green tissues, material-friendly cleaning and disinfection wipes for sensitive surfaces
Catalog # BODE9819430_1
Supplier: BODE CHEMIE
Jump to:
Product Details & Documents
The ready-to-use Bacillol® 30 sensitive green tissues disinfectant wipes from the flowpack, soaked with Bacillol® 30 sensitive foam, wet the surfaces excellently and work quickly, even on sensitive surfaces.
- 100% plastic-free wipes
- Free of microplastics
- Fast-acting with good cleaning properties
- Particularly gentle on materials, also suitable for sensitive materials such as: polycarbonate (PC), acrylic glass (PMMA) and polysulfone
- It can be used without gloves
- Skin-friendly and allergy-friendly
Effectiveness Bactericidal, yeasticidal, tuberculocidal, mycobactericidal, limited virucidal (incl. HBV, HIV, HCV), limited virucidal PLUS.
: SGS Institut Fresenius**- and ECARF-certified.
Specifications
- Description:Bacillol 30 sensitive green tissues, material-friendly cleaning and disinfection wipes for sensitive surfaces
- Seq:02