(S)-10-hydroxycamptothecin
Glass bottle, 5 g
Catalog # 467490050.
Supplier: Thermo Fisher Scientific
(S)-10-hydroxycamptothecin
Catalog # 467490050.
Supplier: Thermo Fisher Scientific
CAS Number: 19685-09-7
Jump to:
Product Details & Documents
Synonyms: 1-HCPT, (S)-4-Ethyl-4,9-dihydroxy-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione, 10-Hydroxycamptothecin
- CAS Number:19685-09-7
- Formula:C₂₀H₁₆N₂O₅
- MDL Number:MFCD02093100
- Molecular Weight:364.35 g/mol
- Storage Temperature:Ambient
Specifications
Specification Test Results
- Appearance:white to yellow to orange powder
- Infrared spectrum:Conforms
- HPLC:≥97.5 %
- Loss on drying:≤5 %
Specifications
- Pack type:Glass bottle
- Pk:5 g



