(S)-10-hydroxycamptothecin
Glass bottle, 1 g
467490010.
: 1-HCPT, (S)-4-Ethyl-4,9-dihydroxy-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione, 10-Hydroxycamptothecin
- :19685-09-7
- :C₂₀H₁₆N₂O₅
- :MFCD02093100
- :364.35 g/mol
- :Ambient
- Appearance:white to yellow to orange powder
- Infrared spectrum:Conforms
- HPLC:≥97.5 %
- Loss on drying:≤5 %
- Pack type:Glass bottle
- Pk:1 g



