1,4-Benzenedimethanethiol
Glass bottle, 5 g
Catalog # 467610050.
Supplier: Thermo Fisher Scientific
1,4-Benzenedimethanethiol
Catalog # 467610050.
Supplier: Thermo Fisher Scientific
CAS Number: 105-09-9
Jump to:
Product Details & Documents
Synonyms: alpha,alpha'-Dimercapto-p-xylene, 1,4-Bis(mercaptomethyl)benzene, 1,4-BDMT
- CAS Number:105-09-9
- Formula:C₈H₁₀S₂
- MDL Number:MFCD00004872
- Molecular Weight:170.29 g/mol
- Boiling Pt:156°C (16hPa)
- Storage Temperature:Ambient
- EINECS:203-269-4
- UN:3335
- ADR:9,III
Specifications
Specification Test Results
- Appearance:white powder, crystals or solid
- Infrared spectrum:Conforms
- GC:≥97.5 %
- Melting point:40°C to 45°C
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Cat. No.:467610050.



