ContactUsLinkComponent
Vacuum Pump V-180. Capacity 1.5m3 /hr. Final vacuum 10mbar, incl. speed contro, including interface I-180 and Woulff bottle, Without Condensation assembly. 1 * 1 items
Catalog # BUCH11V180210
Supplier:  BUCHI
New Product
Vacuum Pump V-180. Capacity 1.5m3 /hr. Final vacuum 10mbar, incl. speed contro, including interface I-180 and Woulff bottle, Without Condensation assembly. 1 * 1 items
Catalog # BUCH11V180210
Supplier:  BUCHI
New Product