Storage Cleaners Cabinets
Cleaners cabinet on casters, 2, Stainless steel 304, 500×800×1800 mm
Catalog # PALBJC0850C
Supplier: Palbam
Jump to:
Product Details & Documents
Designed for storing mops and cleaning materials.
- Two doors
Fabrication material stainless steel 304 with electropolished finish.
Specifications
- Description:Cleaners cabinet on casters
- Material:Stainless steel 304
- Seq:01
- W×D×H:500×800×1800 mm
- Doors:2