Order Entry
Export
Orders LinkContactUsLinkComponent
AMPIGENE® Taq mix

AMPIGENE® Taq mix

AMPIGENE® Taq mix, 1 KIT
Catalog # ENZOENZNUC1001000
Supplier:   Enzo Life Sciences
AMPIGENE® Taq mix
Catalog # ENZOENZNUC1001000
Supplier:  Enzo Life Sciences
CAS Number:  
AMPIGENE® Taq mix

AMPIGENE® Taq mix, 1 KIT

CAS Number:  
Price
609,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents

Optimised standard PCR mix for enhanced speed, yield, and specificity. Used for standard PCR applications.

AMPIGENE® Taq Mix uses the latest developments in polymerase technology and buffer chemistry to optimize PCR. The AMPIGENE® Taq Mix is a robust mix for routine PCR applications including genotyping, screening, and library construction. An optimized buffer system allows efficient amplification under fast and standard cycling conditions.

Delivery information: Kit contains 2×AMPIGENE® Taq Mix (AMPIGENE® Taq Polymerase, 6 mM MgCl₂, 2 mM dNTPs, enhancer and stabilizers).

Specifications

Specifications

  • Pk:
    1 KIT
  • Description:
    AMPIGENE® Taq mix
  • Cat. No.:
    ENZOENZNUC1001000
  • Storage conditions:
    −20 °C

Specifications