Order Entry
Export
Orders LinkContactUsLinkComponent
Antifade mounting medium

Antifade mounting medium

Red antifade reagent, 10 ml, 10 mL
Catalog # ENZOENZ53002M010
Supplier:   Enzo Life Sciences
Antifade mounting medium
Catalog # ENZOENZ53002M010
Supplier:  Enzo Life Sciences
CAS Number:  
Antifade mounting medium

Red antifade reagent, 10 ml, 10 mL

CAS Number:  
Price
170,00 €
Log in for availability
1 * 10 mL
Product Details & Documents

Antifade mounting medium (hardset) is an antifade reagent that causes little or no quenching. Use of the mounting medium significantly extends the photobleaching half-life of the fluorescent stain.

  • Inhibits photobleaching of fluorescent dyes and fluorescent proteins
  • Hardening formulation

Recommended for use on fixed cells and tissues that have been stained with fluorescent stains. Cells can be viewed immediately, or if long term storage is desired, the reagent can be allowed to harden overnight.

Specifications

Specifications

  • Pk:
    10 mL
  • Description:
    Red antifade reagent, 10 ml

Specifications