Order Entry
Export
Orders LinkContactUsLinkComponent
(±)-10-Hydroxycamptothecin ≥97%

(±)-10-Hydroxycamptothecin ≥97%

100 mg
Catalog # ENZOBMLGR3160100
Supplier:   Enzo Life Sciences
(±)-10-Hydroxycamptothecin ≥97%
Catalog # ENZOBMLGR3160100
Supplier:  Enzo Life Sciences
CAS Number:  64439-81-2
(±)-10-Hydroxycamptothecin ≥97%
(±)-10-Hydroxycamptothecin ≥97%

100 mg

CAS Number:  64439-81-2
Price
439,00 €
Log in for availability
1 * 100 mg
Product Details & Documents

A DNA topoisomerase I inhibitor that induces differentiation in HL-60 cells. It induces apoptosis in human breast cancer cell lines MCF-7 and MDA-MB-468.

  • Topoisomerase inhibitor

Pale yellow solid.

Soluble in DMSO.

Synonyms:  1-HCPT, 10-Hydroxycamptothecin, 4-Ethyl-4,9-dihydroxy-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione, NSC107124
  • CAS Number:64439-81-2
  • Formula:C₂₀H₁₆N₂O₅
  • MDL Number:MFCD09027971
  • Molecular Weight:364.35 g/mol
  • Storage Temperature:Freezer
Specifications

Specifications

  • Pk:
    100 mg

Specifications