Order Entry
Export
Orders LinkContactUsLinkComponent
Mitomycin C ≥95%

Mitomycin C ≥95%

10 mg
Catalog # ENZOBMLGR3110010
Supplier:   Enzo Life Sciences
Mitomycin C ≥95%
Catalog # ENZOBMLGR3110010
Supplier:  Enzo Life Sciences
CAS Number:  50-07-7
Mitomycin C ≥95%
Mitomycin C ≥95%

10 mg

CAS Number:  50-07-7
Price
209,00 €
Log in for availability
1 * 10 mg
Product Details & Documents

Antitumor antibiotic. DNA crosslinking and damaging agent. Inhibitor of DNA synthesis, nuclear division and cancer cells. DNA intercalating agent. Induces apoptosis in a variety of cells via a p53-dependent pathway.

  • Blue-violet solid

Soluble in water (1 mg/mL, warm), 100% ethanol (1 mg/mL, warm), DMSO (15 mg/mL) or methanol

Synonyms:  Mitomycin, Ametycin
  • CAS Number:50-07-7
  • Formula:C₁₅H₁₈N₄O₅
  • MDL Number:MFCD00078109
  • Molecular Weight:334.33 g/mol
  • Storage Temperature:Freezer
Specifications

Specifications

  • Pk:
    10 mg

Specifications