Order Entry
Export
Orders LinkContactUsLinkComponent
Wnt pathway library, Screen-Well®

Wnt pathway library, Screen-Well®

Screen-Well® Wnt pathway library, 100 µl/well, 1 SET
Catalog # ENZOBML28380100
Supplier:   Enzo Life Sciences
Wnt pathway library, Screen-Well®
Catalog # ENZOBML28380100
Supplier:  Enzo Life Sciences
CAS Number:  
Wnt pathway library, Screen-Well®

Screen-Well® Wnt pathway library, 100 µl/well, 1 SET

CAS Number:  
Price
1 740,00 €
Log in for availability
1 * 1 SET
Product Details & Documents

The Screen-Well® Wnt pathway library v 2.0 is a focused collection of 71 compounds with defined and diverse Wnt pathway activity, including activators and inhibitors of Wnts, Dishevelled, GSK-3β and other Wnt pathway kinases, TCF/β-catenin, DKK, LRP, Axin, Porcupine, and more. Compounds are dissolved in DMSO at 10 mM or 1 mM and aliquoted into deep-well plates at 100 or 500 µl per well. A variety of structurally and mechanistically different compound classes are included. A useful tool for studying the roles of pro- and anti-Wnt pathway molecules in cells as well as for use in in vitro applications.

The Screen-Well® Wnt Pathway Library v 2.0 is a focused collection of 71 compounds with defined and diverse Wnt pathway activity, including activators and inhibitors of Wnts, Dishevelled, GSK-3β and other Wnt pathway kinases, TCF/β-catenin, DKK, LRP, Axin, Porcupine, and more. Compounds are dissolved in DMSO at 10mM or 1mM and aliquoted into deep-well plates at 100 or 500µL per well. A variety of structurally and mechanistically different compound classes are included. A useful tool for studying the roles of pro- and anti-Wnt pathway molecules in cells as well as for use in in vitro applications.

Ordering information: Contains 71 compounds with defined and diverse Wnt pathway activity. A variety of structurally and mechanistically different compound classes are included.

Specifications

Specifications

  • Pk:
    1 SET
  • Description:
    Screen-Well® Wnt pathway library, 100 µl/well
  • Cat. No.:
    ENZOBML28380100
  • Concentration:
    DMSO solutions (10 mM ) except for Bafilomycin A which is provided at 1 mM in DMSO

Specifications