Order Entry
Export
Orders LinkContactUsLinkComponent
Autophagy library, Screen-Well®

Autophagy library, Screen-Well®

Screen-Well® Autophagy library, 500 µl/well, 1 SET
Catalog # ENZOBML28370500
Supplier:   Enzo Life Sciences
Autophagy library, Screen-Well®
Catalog # ENZOBML28370500
Supplier:  Enzo Life Sciences
CAS Number:  
Autophagy library, Screen-Well®

Screen-Well® Autophagy library, 500 µl/well, 1 SET

CAS Number:  
Price
3 130,00 €
Log in for availability
1 * 1 SET
Product Details & Documents

Structurally and mechanistically different compounds for studying the roles of pro- and anti-autophagic molecules in cells and in vitro applications.

The Screen-well® autophagy library contains 94 compounds with defined autophagy-inducing or -inhibitory activity. Compounds are dissolved in DMSO at 10 mM or 1 mM and aliquoted into deep-well plates at 100 or 500 µl per well. A variety of structurally and mechanistically different compound classes are included. A useful tool for studying the roles of pro- and anti-autophagic molecules in cells as well as for use in in vitro applications.

Ordering information: Contains 94 compounds with defined autophagy-inducing or -inhibitory activity.  A variety of structurally and mechanistically different compound classes are included.

Specifications

Specifications

  • Pk:
    1 SET
  • Description:
    Screen-Well® Autophagy library, 500 µl/well
  • Cat. No.:
    ENZOBML28370500
  • Concentration:
    DMSO solutions (10 mM ) except for Bafilomycin A which is provided at 1 mM in DMSO.

Specifications