Orders LinkContactUsLinkComponent
Epigenetics library, Screen-Well®
Epigenetics library, Screen-Well®
Screen-Well® Epigenetics library, 100 µl/well, 1 SET
Catalog # ENZOBML28360100
Supplier:   Enzo Life Sciences
Epigenetics library, Screen-Well®
Catalog # ENZOBML28360100
Supplier:  Enzo Life Sciences
CAS Number:  
Epigenetics library, Screen-Well®
Screen-Well® Epigenetics library, 100 µl/well, 1 SET
CAS Number:  
Price
1 620,00 €
Log in for availability
1 * 1 SET
Product Details & Documents
The Screen-Well® epigenetics library contains 43 compounds with defined activity against enzymes which carry out epigenetic modification of lysine. It also includes DNA methylation inhibitors. Compounds are dissolved in DMSO at 10 mM and aliquoted into deep-well plates at 100 µl or 500 µl per well. The library is a useful tool for chemical genomics, assay development and other pharmacological applications. The library contains inhibitors of these important enzymes: HDACs, SIRTs, Lysine demethylases, HATs, Histone methyl transferases, DNA methyltransferases as well as SIRT activators. A variety of structurally and mechanistically different compound classes are included.
The Screen-Well® Epigenetics library contains 43 compounds with defined activity against enzymes which carry out epigenetic modification of lysine. It also includes DNA methylation inhibitors. Compounds are dissolved in DMSO at 10mM and aliquoted into deep-well plates at 100 or 500µL per well. The library is a useful tool for chemical genomics, assay development and other pharmacological applications. The library contains inhibitors of these important enzymes: HDACs, SIRTs, Lysine demethylases, HATs, Histone methyl transferases, DNA methyltransferases as well as SIRT activators. A variety of structurally and mechanistically different compound classes are included.
Ordering information: Contains 43 compounds supplied in DMSO at a concentration of 10mM.
Specifications
Specifications
  • Pk:
    1 SET
  • Description:
    Screen-Well® Epigenetics library, 100 µl/well
Specifications