Order Entry
Export
Orders LinkContactUsLinkComponent
Cycloheximide ≥95% (by TLC)

Cycloheximide ≥95% (by TLC)

5 g
Catalog # ENZOALX380269G005
Supplier:   Enzo Life Sciences
Cycloheximide ≥95% (by TLC)
Catalog # ENZOALX380269G005
Supplier:  Enzo Life Sciences
CAS Number:  66-81-9
Cycloheximide ≥95% (by TLC)

5 g

CAS Number:  66-81-9
Price
250,00 €
Log in for availability
1 * 5 g
Product Details & Documents

Protein synthesis inhibitor

Antibiotic. Inhibits protein synthesis in eukaryotes by interfering with the translocation step. Induces apoptosis in tumor cell lines but can also delay or inhibit apoptosis induced by other agents. Widely used for selection of resistant strains of yeast and fungy, controlled inhibiton of protein synthesis for detecton of short-lived proteins and super-induction of protein expression. At 10µg/ml cycloheximide totally inhibits protein synthesis in mouse thymocytes.

Isolated from Streptomyces griseus.

Synonyms:  Naramycin A, Actidione, 3-[2-(3,5-Dimethyl-2-oxocyclohexyl)-2-hydroxyethyl]glutarimide,
  • CAS Number:66-81-9
  • Formula:C₁₅H₂₃NO₄
  • MDL Number:MFCD00082346
  • Molecular Weight:281.35 g/mol
Specifications

Specifications

  • Pk:
    5 g

Specifications