G418 disulphate
Glass bottle for solids, 500 mg
Catalog # ENZOALX380013M500
Supplier: Enzo Life Sciences
Jump to:
Product Details & Documents
Synonyms: G-418 sulphate, Geneticin®, G-418 disulphate, G418 sulphate
- CAS Number:108321-42-2
- Formula:C₂₀H₄₄N₄O₁₈S₂
- MDL Number:MFCD00058314
- Molecular Weight:692.72 g/mol
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:500 mg



