Order Entry
Export
Orders LinkContactUsLinkComponent
G418 disulphate

G418 disulphate

Glass bottle for solids, 5 g
Catalog # ENZOALX380013G005
Irritant
Supplier:   Enzo Life Sciences
G418 disulphate
Catalog # ENZOALX380013G005
Supplier:  Enzo Life Sciences
CAS Number:  108321-42-2
G418 disulphate
G418 disulphate

Glass bottle for solids, 5 g

CAS Number:  108321-42-2
Price
272,00 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  G-418 sulphate, Geneticin®, G-418 disulphate, G418 sulphate
  • CAS Number:108321-42-2
  • Formula:C₂₀H₄₄N₄O₁₈S₂
  • MDL Number:MFCD00058314
  • Molecular Weight:692.72 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications