Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
G418 disulphate
G418 disulphate
G418 disulphate
Glass bottle for solids, 1 g
ENZOALX380013G001
:
Enzo Life Sciences
G418 disulphate
ENZOALX380013G001
:
Enzo Life Sciences
:
108321-42-2
G418 disulphate
Glass bottle for solids, 1 g
:
108321-42-2
71,50 €
1 * 1 g
-
+
:
G-418 sulphate, Geneticin®, G-418 disulphate, G418 sulphate
:
108321-42-2
:
C₂₀H₄₄N₄O₁₈S₂
:
MFCD00058314
:
692.72 g/mol
:
Ambient
Pack type:
Glass bottle for solids
Pk:
1 g