Order Entry
Export
Orders LinkContactUsLinkComponent
Gentamicin sulphate

Gentamicin sulphate

Glass bottle for solids, 10 g
Catalog # ENZOALX380003G010
Health
Supplier:   Enzo Life Sciences
Gentamicin sulphate
Catalog # ENZOALX380003G010
Supplier:  Enzo Life Sciences
CAS Number:  1405-41-0
Gentamicin sulphate

Glass bottle for solids, 10 g

CAS Number:  1405-41-0
Price
522,00 €
Log in for availability
1 * 10 g
Product Details & Documents
Synonyms:  NMD Inhibitor II, Garamycin, Gentamicin sulphate salt, Gentiomycin C
  • CAS Number:1405-41-0
  • MDL Number:MFCD00270181
  • Merck Index:13,04403
  • Melting Pt:218…237 °C
  • Storage Temperature:Ambient
  • EINECS:215-778-9
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g

Specifications