Order Entry
Export
Orders LinkContactUsLinkComponent
 

trans-Ned-19

0,5 mg
Catalog # ENZOALX270503MC05
Supplier:   Enzo Life Sciences
trans-Ned-19
Catalog # ENZOALX270503MC05
Supplier:  Enzo Life Sciences
CAS Number:  1354235-96-3
trans-Ned-19

0,5 mg

CAS Number:  1354235-96-3
Price
267,00 €
Log in for availability
1 * 0,5 mg
Product Details & Documents

Cell permeable, selective and potent inhibitor of nicotinic acid adenine dinucleotide phosphate (NAADP) receptor in mammalian cells and sea urchin eggs. Blocks NAADP signaling. Inhibits Ca2+ release: trans-Ned-19 (IC50 = 6nM); cis-Ned-19 (IC50 = 800nM). [32P] NAADP binding: trans-Ned-19 (IC50 = 0.4nM); cis-Ned-19 (IC50 = 15µM).
Inhibits Ca2+ release from lysosomes. Noncompetitive antagonist. Fluorescent.

Synthetic

  • CAS Number:1354235-96-3
  • Formula:C₃₀H₃₁FN₄O₃
  • MDL Number:MFCD02057367
  • Molecular Weight:514.601 g/mol
Specifications

Specifications

  • Pk:
    0,5 mg

Specifications