Order Entry
Export
Orders LinkContactUsLinkComponent
Ras inhibitory peptide

Ras inhibitory peptide

1 mg
Catalog # ENZOALX166001M001
Supplier:   Enzo Life Sciences
Ras inhibitory peptide
Catalog # ENZOALX166001M001
Supplier:  Enzo Life Sciences
CAS Number:  
Ras inhibitory peptide

1 mg

CAS Number:  
Price
200,00 €
Log in for availability
1 * 1 mg
Product Details & Documents

The peptide sequence corresponds to amino acid residues 1149-1158 of the guanine nucleotide-releasing factor hSos1, which is essential for the control of Ras activity. Blocks the binding of hSos1 to human Grb2, a protein that binds to activated receptor tyrosine kinases.

Specifications

Specifications

  • Pk:
    1 mg
  • Cat. No.:
    ENZOALX166001M001
  • Protein/peptide name:
    Ras inhibitory Peptide
  • Purity:
    >97%

Specifications