Order Entry
Export
Orders LinkContactUsLinkComponent
L(-)-Glutathione (reduced form)

L(-)-Glutathione (reduced form)

Glass bottle for solids, 25 g
Catalog # ENZOALX157002G025
Supplier:   Enzo Life Sciences
L(-)-Glutathione (reduced form)
Catalog # ENZOALX157002G025
Supplier:  Enzo Life Sciences
CAS Number:  70-18-8
L(-)-Glutathione (reduced form)

Glass bottle for solids, 25 g

CAS Number:  70-18-8
Price
178,00 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  L-Glutathione Reduced, GSH, (S)-2-Amino-5-(((R)-1-((carboxymethyl)amino)-3-mercapto-1-oxopropan-2-yl)amino)-5-oxopentanoic acid, Glutathione Reduced
  • CAS Number:70-18-8
  • Formula:C₁₀H₁₇N₃O₆S
  • MDL Number:MFCD00065939
  • Molecular Weight:307.33 g/mol
  • Merck Index:13,04488
  • Melting Pt:185…195 °C
  • Density:1,475 g/cm³ (20 °C)
  • Storage Temperature:Refrigerator
  • EINECS:200-725-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications