Order Entry
Export
Orders LinkContactUsLinkComponent
Lysates, HeLa (human)

Lysates, HeLa (human)

Lysate, HeLa, human, heat shocked, Pooled, 200 µG
Catalog # ENZOADILYCHL101F
Supplier:   Enzo Life Sciences
Lysates, HeLa (human)
Catalog # ENZOADILYCHL101F
Supplier:  Enzo Life Sciences
CAS Number:  
Lysates, HeLa (human)
Lysates, HeLa (human)

Lysate, HeLa, human, heat shocked, Pooled, 200 µG

CAS Number:  
Price
170,00 €
Log in for availability
1 * 200 µG
Product Details & Documents

The HeLa cell line derived from a human donor's epithelioid carcinoma tissue (cervix).

Dilution range: 20 µg/lane (Western blotting)

Liquid lysates in SDS-PAGE sample buffer.

Heat shock-treated HeLa cells with no recovery time can be a useful tool in studying the regulators of immediate-early gene expression during heat shock stress response.

Specifications

Specifications

  • Pk:
    200 µG
  • Single Donor/Pooled:
    Pooled
  • Description:
    Lysate, HeLa, human, heat shocked

Specifications