Aldosterone ELISA kit
Aldosterone ELISA kit, 1×96 wells, 1 KIT
Catalog # ENZOADI900173
Supplier: Enzo Life Sciences
Jump to:
Product Details & Documents
The Aldosterone EIA kit is a colorimetric competitive enzyme immunoassay kit for the quantitative determination of aldosterone in plasma, serum, and urine from any species.
- Highly sensitive
- Negligible reactivity with other steroids
- Fully quantitative results that surpass semi-quantitative Western blots
- Results overnight + 1 hour
Delivery information: Kit includes DxS microtiter plate, conjugate, antibody, assay buffer concentrate, wash buffer concentrate, standard, pNpp substrate, stop solution.
Specifications
Specifications
- Pk:1 KIT
- Description:Aldosterone ELISA kit
- Tests per kit:1×96 wells
- Cat. No.:ENZOADI900173
- Sensitivity:4,7 pg/ml (range 3,9 - 250 pg/ml)
- Conjugate ELISA:Alkaline Phosphatase (AP)
- Format:Pre-coated
- Primary antibody reactivity:Species independent
- Sample type:Plasma, serum, and urine samples
- Target protein:Aldosterone
- Storage conditions:–20 °C (long term)
- Assay Type:Competitive



