Orders LinkContactUsLinkComponent
Cleanroom ESD Ergo Chairs
Cleanroom ESD Ergo Chairs
Cleanroom chair with flat conductive glides
Catalog # PALBPCKD0120
Supplier:   Palbam
Cleanroom ESD Ergo Chairs
Catalog # PALBPCKD0120
Supplier:  Palbam
Cleanroom ESD Ergo Chairs
Cleanroom ESD Ergo Chairs
Cleanroom chair with flat conductive glides
Price
2 400,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Cleanroom ESD ergo chair which lives up to our reputation for high quality and ergonomics/operator comfort.
  • Seat and backrest material and colour: Polyurethane (PU), Black (RAL 9011)
  • Base Material: Chrome steel
Seat height adjustable from 450 to 640 mm.
The chairs include the 'automatic weight adjustment' ergo feature and a tapering backrest offering optimum freedom for shoulders. Options available for conductive glides or casters.
Certifications: The chairs comply with: Cleanliness class 4 DIN EN ISO 14644-1 Anti-static and conductive according to DIN EN 61340-5-1:2000.
Specifications
  • Description:
    Cleanroom chair with flat conductive glides
  • Seq:
    03