Supports for stand grids
Flat flanges LF 100, framework grid 0,500 m
Catalog # LENZ06247211
Supplier: Lenz Laborglas
Jump to:
Product Details & Documents
Stainless steel (clamp), steel, powder coated (supports).
For fixing lids onto reaction vessels and for the suspension into stands, complete with clamp and supports.
Specifications
- For:Flat flanges LF 100, framework grid 0,500 m
- Seq:01