Ammonia solution 27 - 30% FCC
Glass bottle, poly-coated, 4 L
SPCMA1293-4LTPV
Ammonia Solution, FCC is a solution of Ammonia in water. It has many uses, but in food production it is used primarily as a leavening agent. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
GHS hazard : Corrosion;Exclamation mark
UN2672
: Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
- :1336-21-6
- :NH₃
- :MFCD00066650
- :35.05 g/mol
- :0,900
- :Ambient
- :2672
- :8,III
- Assay (NH₃, By Wt.):27.0 - 30.0%
- Lead (Pb):max. 0.5 mg/kg
- Nonvolatile Residue:max. 0.02 %
- Readily Oxidizable Substances:To Pass Test
- Identification:To Pass Test
- Pack type:Glass bottle, poly-coated
- Pk:4 L



