Order Entry
Export
Orders LinkContactUsLinkComponent
2,6-Dimethyl-4-nitroaniline 95%

2,6-Dimethyl-4-nitroaniline 95%

Bottle, 25 g
Catalog # APOSOR62166-25G
Irritant
Supplier:   Apollo Scientific
2,6-Dimethyl-4-nitroaniline 95%
Catalog # APOSOR62166-25G
Supplier:  Apollo Scientific
CAS Number:  16947-63-0
2,6-Dimethyl-4-nitroaniline 95%

Bottle, 25 g

CAS Number:  16947-63-0
Price
973,00 €
Log in for availability
1 * 25 g
Product Details & Documents
  • CAS Number:16947-63-0
  • Formula:C₈H₁₀N₂O₂
  • MDL Number:MFCD00211068
  • Molecular Weight:166.18 g/mol
  • Melting Pt:165 °C
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g

Specifications