Order Entry
Export
Orders LinkContactUsLinkComponent
3,4-Dihydroxybenzaldehyde

3,4-Dihydroxybenzaldehyde

Bottle, 500 g
Catalog # APOSOR5940-500G
Irritant
Supplier:   Apollo Scientific
3,4-Dihydroxybenzaldehyde
Catalog # APOSOR5940-500G
Supplier:  Apollo Scientific
CAS Number:  139-85-5
3,4-Dihydroxybenzaldehyde

Bottle, 500 g

CAS Number:  139-85-5
Price
167,00 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Protocatechuic aldehyde, Protocatechualdehyde
  • CAS Number:139-85-5
  • Formula:C₇H₆O₃
  • MDL Number:MFCD00003370
  • Molecular Weight:138.12 g/mol
  • Melting Pt:150…157 °C (dec.)
  • Storage Temperature:Ambient
  • EINECS:205-377-7
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 g

Specifications