Order Entry
Export
Orders LinkContactUsLinkComponent
3,4-Dichlorophenylacetylene

3,4-Dichlorophenylacetylene

Bottle, 25 g
Catalog # APOSOR5185-25G
Irritant
Supplier:   Apollo Scientific
3,4-Dichlorophenylacetylene
Catalog # APOSOR5185-25G
Supplier:  Apollo Scientific
CAS Number:  556112-20-0
3,4-Dichlorophenylacetylene

Bottle, 25 g

CAS Number:  556112-20-0
Price
278,00 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  1,2-Dichloro-4-ethynylbenzene
  • CAS Number:556112-20-0
  • Formula:C₈H₄Cl₂
  • MDL Number:MFCD07783046
  • Molecular Weight:171.023 g/mol
  • Melting Pt:40…44 °C
  • Flash Pt:96.1 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g

Specifications