Order Entry
Export
Orders LinkContactUsLinkComponent
 

1,1-Dichloroethane 98%

Bottle, 1 g
Catalog # APOSOR3712-1G
Flammable
Irritant
Supplier:   Apollo Scientific
1,1-Dichloroethane 98%
Catalog # APOSOR3712-1G
Supplier:  Apollo Scientific
CAS Number:  75-34-3
1,1-Dichloroethane 98%

Bottle, 1 g

CAS Number:  75-34-3
Price
109,00 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  Ethylidene dichloride
  • CAS Number:75-34-3
  • Formula:Cl₂HCCH₃
  • MDL Number:MFCD00013673
  • Molecular Weight:98.96 g/mol
  • Boiling Pt:57 °C
  • Melting Pt:–98 °C
  • Density:1.18 g/cm³ (25 °C)
  • Flash Pt:–10 °C
  • Storage Temperature:Ambient
  • EINECS:200-863-5
  • UN:2362
  • ADR:3,II
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications