Order Entry
Export
Orders LinkContactUsLinkComponent
ω,2,4-Trichloroacetophenone

ω,2,4-Trichloroacetophenone

Bottle, 1 kg
Catalog # APOSOR18371-1KG
Corrosive
Toxic
Supplier:   Apollo Scientific
ω,2,4-Trichloroacetophenone
Catalog # APOSOR18371-1KG
Supplier:  Apollo Scientific
CAS Number:  4252-78-2
ω,2,4-Trichloroacetophenone

Bottle, 1 kg

CAS Number:  4252-78-2
Price
152,00 €
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  2,4-Dichlorophenacyl chloride, α,2,4-Trichloroacetophenone, 2,2',4'-Trichloroacetophenone
  • CAS Number:4252-78-2
  • Formula:C₈H₅Cl₃O
  • MDL Number:MFCD00000934
  • Molecular Weight:223.49 g/mol
  • Boiling Pt:130…135 °C/4mmHg
  • Melting Pt:47…56 °C
  • Density:1.312 g/cm³ (25 °C)
  • Flash Pt:>110 °C
  • Storage Temperature:Ambient
  • EINECS:224-218-2
  • UN:2923
  • ADR:8+6.1,III
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 kg

Specifications